siRNA Details
Gene Symbol: Ssfa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080558.4 | NP_542125.3 | 18 | 4631 | ||
GenBank mRNA | AB093303.1 | - | 18 | 4506 | ||
AK037213.1 | - | 16 | 4341 | |||
AK169543.1 | - | 19 | 4809 | |||
BC008560.1 | - | 11 | 3604 | |||
BC018526.1 | - | 8 | 1835 | |||
BC079535.1 | - | 12 | 4010 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
