siRNA Details
Gene Symbol: FCGR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127593.1 | NP_001121065.1 | 6 | 1029 | ||
NM_001127595.1 | NP_001121067.1 | 6 | 1011 | |||
NM_001127596.1 | NP_001121068.1 | 6 | 1008 | |||
XM_011509293.2 | XP_011507595.2 | 4 | 660 | |||
GenBank mRNA | AK223268.1 | - | 5 | 890 | ||
AK223295.1 | - | 5 | 902 | |||
AK302126.1 | - | 5 | 1241 | |||
BC017865.1 | - | 5 | 922 | 4690249 | ||
BC033678.1 | - | 5 | 897 | 5180561 | ||
BC036723.1 | - | 5 | 1166 | 5206097 | ||
M24853.1 | - | 3 | 784 |
Gene Symbol: FCGR3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000570.4 | NP_000561.3 | 6 | 996 | ||
NM_001244753.1 | NP_001231682.1 | 5 | 1242 | |||
NM_001271035.1 | NP_001257964.1 | 5 | 1239 | |||
NM_001271036.1 | NP_001257965.1 | 5 | 1108 | |||
NM_001271037.1 | NP_001257966.1 | 4 | 850 | |||
GenBank mRNA | AJ581669.1 | - | 5 | 907 | ||
J04162.1 | - | 5 | 891 | |||
M24854.1 | - | 5 | 972 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
