siRNA Details
Gene Symbol: Nek8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080849.3 | NP_543125.1 | 15 | 2750 | ||
GenBank mRNA | AF407579.1 | - | 15 | 2706 | ||
AK154358.1 | - | 15 | 2713 | |||
BC057995.1 | - | 14 | 2832 | |||
BC070457.1 | - | 15 | 2699 | 30603080 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
