siRNA Details
Gene Symbol: Dusp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048054.2 | NP_001041519.1 | 7 | 3268 | ||
NM_130447.3 | NP_569714.2 | 7 | 4234 | |||
GenBank mRNA | AB052157.1 | - | 7 | 3948 | ||
AF345951.1 | - | 7 | 4072 | |||
AF345952.1 | - | 6 | 3907 | |||
AF345953.1 | - | 7 | 3106 | |||
AF345954.1 | - | 6 | 2941 | |||
AK014441.1 | - | 1 | 1707 | |||
BC057321.1 | - | 7 | 4085 | 6830600 | ||
BC059232.1 | - | 7 | 4022 | 6409944 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
