siRNA Details
Gene Symbol: Card14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130886.3 | NP_570956.1 | 19 | 2868 | ||
GenBank mRNA | AF363457.1 | - | 19 | 2847 | ||
AK028888.1 | - | 13 | 2253 | |||
BC004692.1 | - | 15 | 2239 | |||
BC029102.1 | - | 19 | 2868 | 3979883 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
