siRNA Details
Gene Symbol: C4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002029.3 | NP_001002029.3 | 26 | 3413 | ||
GenBank mRNA | AK304564.1 | - | 5 | 528 | ||
K02404.1 | - | 4 | 497 |
Gene Symbol: C4B_2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242823.2 | NP_001229752.1 | 26 | 3413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
