siRNA Details
Gene Symbol: Rac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133223.4 | NP_573486.1 | 3 | 330 | ||
GenBank mRNA | AB040819.1 | - | 3 | 197 | ||
AK089930.1 | - | 3 | 331 | |||
AK150203.1 | - | 3 | 335 | |||
AY974601.1 | - | 2 | 169 | |||
BC025842.1 | - | 2 | 202 | |||
BC117028.1 | - | 3 | 328 | 40126347 | ||
BC119041.1 | - | 3 | 328 | 8733790 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
