siRNA Details
Gene Symbol: Atad3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_179203.3 | NP_849534.2 | 1 | 232 | ||
GenBank mRNA | AF343079.1 | - | 1 | 278 | ||
AK075921.1 | - | 1 | 228 | |||
AK148974.1 | - | 1 | 231 | |||
AK149702.1 | - | 1 | 231 | |||
AK150037.1 | - | 1 | 231 | |||
AK165724.1 | - | 1 | 229 | |||
AK166099.1 | - | 1 | 228 | |||
AK172038.1 | - | 1 | 228 | |||
AK173141.1 | - | 1 | 180 | |||
BB839884.1 | - | 1 | 233 | |||
BC023301.1 | - | 1 | 169 | 3671787 | ||
BC047047.1 | - | 1 | 195 | |||
BC058373.1 | - | 1 | 216 | 6828037 | ||
BC060036.1 | - | 1 | 164 | 6530155 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001034922, BC105762.
Gene Ontology Categories:
