siRNA Details
Gene Symbol: Atad3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_179203.3 | NP_849534.2 | 2, 3 | 382 | ||
GenBank mRNA | AF343079.1 | - | 2, 3 | 428 | ||
AK075921.1 | - | 2, 3 | 378 | |||
AK148974.1 | - | 2, 3 | 381 | |||
AK149702.1 | - | 2, 3 | 381 | |||
AK150037.1 | - | 2, 3 | 381 | |||
AK165724.1 | - | 2, 3 | 379 | |||
AK166099.1 | - | 2, 3 | 378 | |||
AK172038.1 | - | 2, 3 | 378 | |||
AK172350.1 | - | 2, 3 | 383 | |||
AK173141.1 | - | 2, 3 | 330 | |||
BB839884.1 | - | 2, 3 | 383 | |||
BC023301.1 | - | 2, 3 | 319 | 3671787 | ||
BC047047.1 | - | 2, 3 | 345 | |||
BC058373.1 | - | 2, 3 | 366 | 6828037 | ||
BC060036.1 | - | 2, 3 | 314 | 6530155 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
