siRNA Details
Gene Symbol: Apcdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB023957.1 | - | 314 | |||
AK157407.1 | - | 1575 | ||||
AK172004.1 | - | 3012 | ||||
BC065780.1 | - | 287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
