siRNA Details
Gene Symbol: TWF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007284.3 | NP_009215.1 | 4 | 489 | ||
GenBank mRNA | AF109365.1 | - | 4 | 390 | ||
AL136773.1 | - | 4 | 456 | |||
BC000327.2 | - | 4 | 440 | 2822004 | ||
BC003161.1 | - | 4 | 406 | 2957960 | ||
BC016452.1 | - | 4 | 456 | 3452300 | ||
CR533520.1 | - | 4 | 338 | |||
Y17169.1 | - | 4 | 442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
