siRNA Details
Gene Symbol: Zbtb11os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006543.1 | - | 1 | 1109 | ||
AK010004.1 | - | 1 | 1286 | |||
BC017620.1 | - | 1 | 1245 | 4924337 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_133677.1 | - | 1245 | Chr. - - [-] on Build |
