siRNA Details
Gene Symbol: 4931406C07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199484.1 | NP_001186413.1 | 3, 4 | 489 | ||
NM_001199485.1 | NP_001186414.1 | 3, 4 | 385 | |||
NM_133732.3 | NP_598493.1 | 2, 3 | 348 | |||
GenBank mRNA | AK088123.1 | - | 3, 4 | 349 | ||
AK133192.1 | - | 3, 4 | 490 | |||
AK147073.1 | - | 3, 4 | 359 | |||
BC011540.1 | - | 2, 3 | 251 | 2648220 | ||
BC016078.1 | - | 2, 3 | 297 | 4489072 | ||
BC025213.1 | - | 2, 3 | 249 | |||
CT010296.1 | - | 2, 3 | 162 |
