siRNA Details
Gene Symbol: BRD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007371.3 | NP_031397.1 | 12 | 2478 | ||
XM_006717291.2 | XP_006717354.1 | 12 | 2610 | |||
XM_011519052.2 | XP_011517354.1 | 12 | 2829 | |||
XM_017015165.1 | XP_016870654.1 | 11 | 2400 | |||
XM_017015166.1 | XP_016870655.1 | 9 | 1795 | |||
GenBank mRNA | AK299519.1 | - | 12 | 2513 | ||
AY513270.1 | - | 11 | 2308 | |||
D26362.1 | - | 12 | 2429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
