siRNA Details
Gene Symbol: Ugt2b38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133894.2 | NP_598655.1 | 2 | 799 | ||
GenBank mRNA | AK034801.1 | - | 2 | 800 | ||
BC015272.1 | - | 2 | 796 | 4241480 | ||
BC057169.1 | - | 2 | 785 | 4975249 | ||
BC069923.1 | - | 2 | 787 | 4974994 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
