siRNA Details
Gene Symbol: Chpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133913.2 | NP_598674.2 | 1 | 1407 | ||
GenBank mRNA | AK089404.1 | - | 1 | 1417 | ||
AK122504.1 | - | 1 | 1407 | |||
AK138814.1 | - | 1413 | ||||
BC047275.1 | - | 1 | 630 | 6441739 |

More Information
Other Species Targeted

Gene Ontology Categories:
