siRNA Details
Gene Symbol: Nfxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133921.2 | NP_598682.2 | 17, 18 | 2167 | ||
GenBank mRNA | AK132320.1 | - | 16, 17 | 2077 | ||
BC022652.1 | - | 1, 2 | 167 | |||
CF916554.1 | - | 4, 5 | 419 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
