siRNA Details
Gene Symbol: Ttll3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142732.1 | NP_001136204.1 | 3 | 506 | ||
NM_133923.6 | NP_598684.4 | 3 | 623 | |||
GenBank mRNA | AK029462.1 | - | 3 | 507 | ||
AM690746.1 | - | 3 | 623 | |||
BC006830.1 | - | 2 | 149 | |||
BC021404.1 | - | 2 | 255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
