siRNA Details
Gene Symbol: Cops8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133805.3 | NP_598566.3 | 5, 6 | 572 | ||
GenBank mRNA | AF482000.1 | - | 5, 6 | 1033 | ||
AK079111.1 | - | 5, 6 | 506 | |||
AK079522.1 | - | 5, 6 | 520 | |||
AK088323.1 | - | 5, 6 | 492 | |||
AK090013.1 | - | 5, 6 | 501 | |||
AK142989.1 | - | 4, 5 | 1541 | |||
AK145899.1 | - | 5, 6 | 499 | |||
AK162502.1 | - | 5, 6 | 507 | |||
AK195692.1 | - | 4, 5 | 347 | |||
BC017690.1 | - | 5, 6 | 540 | 4238947 | ||
BC021488.1 | - | 5, 6 | 531 | 5344864 | ||
BC024421.1 | - | 5, 6 | 525 | 4987584 |
