siRNA Details
Gene Symbol: BRD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007371.3 | NP_031397.1 | 3 | 505 | ||
XM_006717291.2 | XP_006717354.1 | 3 | 637 | |||
XM_011519052.2 | XP_011517354.1 | 3 | 856 | |||
XM_017015165.1 | XP_016870654.1 | 2 | 430 | |||
GenBank mRNA | AK299519.1 | - | 3 | 637 | ||
AY513270.1 | - | 2 | 335 | |||
BC032124.2 | - | 3 | 615 | 4856840 | ||
BI545861.1 | - | 3 | 515 | |||
D26362.1 | - | 3 | 456 | |||
KR633047.1 | - | 2 | 317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
