siRNA Details
Gene Symbol: Golga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080968.1 | NP_001074437.1 | 16 | 1334 | ||
NM_133852.2 | NP_598613.2 | 17 | 1415 | |||
GenBank mRNA | AK076370.1 | - | 11 | 1033 | ||
AK077516.1 | - | 16 | 1336 | |||
AK220509.1 | - | 17 | 1343 | |||
BC011407.1 | - | 15 | 1135 | 3596079 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
