siRNA Details
Gene Symbol: PLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000533.4 | NP_000524.3 | 3 | 593 | ||
NM_001128834.2 | NP_001122306.1 | 4 | 675 | |||
NM_001305004.1 | NP_001291933.1 | 3 | 368 | |||
GenBank mRNA | AK128782.1 | - | 3 | 516 | ||
AK292728.1 | - | 4 | 675 | |||
AK295374.1 | - | 4 | 446 | |||
AK295388.1 | - | 3 | 347 | |||
AK308966.1 | - | 2 | 322 | |||
AK309239.1 | - | 3 | 534 | |||
AK312340.1 | - | 3 | 516 | |||
BC095452.1 | - | 3 | 477 | 30717624 | ||
BX445448.2 | - | 3 | 494 | |||
KU178262.1 | - | 2 | 390 | |||
M27110.1 | - | 3 | 508 | |||
M54927.1 | - | 3 | 511 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
