siRNA Details
Gene Symbol: Chpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133913.2 | NP_598674.2 | 1 | 1486 | ||
GenBank mRNA | AK089404.1 | - | 1 | 1496 | ||
AK122504.1 | - | 1 | 1486 | |||
AK138814.1 | - | 1492 | ||||
BC047275.1 | - | 1 | 709 | 6441739 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
