siRNA Details
Gene Symbol: Aff1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080798.1 | NP_001074267.1 | 20 | 7860 | ||
NM_133919.3 | NP_598680.3 | 20 | 7871 | |||
GenBank mRNA | AK030884.1 | - | 1 | 1719 | ||
BC018434.1 | - | 1 | 370 | |||
BC085290.1 | - | 2 | 480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
