siRNA Details
Gene Symbol: Nfxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133921.2 | NP_598682.2 | 20 | 2473 | ||
GenBank mRNA | AK132320.1 | - | 19 | 2383 | ||
BC022652.1 | - | 4 | 473 |

More Information
Other Species Targeted

Gene Ontology Categories:
