siRNA Details
Gene Symbol: AU040320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133886.2 | NP_598647.1 | 21 | 3612 | ||
GenBank mRNA | AK147569.1 | - | 6143 | |||
AK161493.1 | - | 11 | 1675 | |||
AK170261.1 | - | 21 | 3569 | |||
BC022154.1 | - | 12 | 2024 | |||
BC028869.1 | - | 21 | 3582 | 4235813 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
