siRNA Details
Gene Symbol: Usb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133954.2 | NP_598715.2 | 2 | 275 | ||
GenBank mRNA | AK135527.1 | - | 2 | 268 | ||
AK160901.1 | - | 2 | 258 | |||
AK168683.1 | - | 2 | 257 | |||
AK186661.1 | - | 2 | 145 | |||
BC016418.1 | - | 2 | 219 | 4224590 |
