siRNA Details
Gene Symbol: Dusp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037955.4 | NP_001033044.1 | 3 | 158 | ||
NM_134068.3 | NP_598829.1 | 3 | 158 | |||
GenBank mRNA | AF237619.1 | - | 3 | 118 | ||
AK148708.1 | - | 3 | 126 | |||
BC057944.1 | - | 3 | 128 | |||
BC080718.1 | - | 3 | 108 | |||
BC108362.1 | - | 3 | 108 | 4917645 | ||
CX236611.1 | - | 3 | 158 |

More Information
Other Species Targeted

Gene Ontology Categories:
