siRNA Details
Gene Symbol: Usb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133954.2 | NP_598715.2 | 5 | 674 | ||
GenBank mRNA | AK135527.1 | - | 5 | 667 | ||
AK160901.1 | - | 5 | 657 | |||
AK168683.1 | - | 5 | 656 | |||
BC016418.1 | - | 5 | 618 | 4224590 |
