siRNA Details
Gene Symbol: Slc22a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133980.3 | NP_598741.2 | 6 | 1049 | ||
GenBank mRNA | AF529220.1 | - | 6 | 1042 | ||
BC021449.1 | - | 6 | 1041 | 5044564 |

More Information
Other Species Targeted

Gene Ontology Categories:
