siRNA Details
Gene Symbol: CXCL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000609.6 | NP_000600.1 | 3, 4 | 369 | ||
GenBank mRNA | AK124641.1 | - | 1 | 1440 | ||
AL713778.1 | - | 1 | 1229 | |||
BX647204.1 | - | 1 | 2171 | |||
L36033.1 | - | 3, 4 | 356 | |||
U16752.1 | - | 3, 4 | 357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
