siRNA Details
Gene Symbol: Vipas39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142580.1 | NP_001136052.1 | 20 | 2092 | ||
NM_001142581.1 | NP_001136053.1 | 19 | 2035 | |||
NM_134044.3 | NP_598805.2 | 19 | 2049 | |||
GenBank mRNA | AK046322.1 | - | 20 | 2063 | ||
AK049643.1 | - | 20 | 2039 | |||
AK079055.1 | - | 20 | 2039 | |||
AK082261.1 | - | 20 | 2020 | |||
AK145043.1 | - | 2076 | ||||
BC016646.1 | - | 19 | 1958 | 2651233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
