siRNA Details
Gene Symbol: Dock9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081039.1 | NP_001074508.1 | 47, 48 | 5418 | ||
NM_001128307.1 | NP_001121779.1 | 46, 47 | 5679 | |||
NM_001128308.1 | NP_001121780.1 | 46, 47 | 5676 | |||
NM_134074.2 | NP_598835.2 | 46, 47 | 5268 | |||
GenBank mRNA | AK045750.1 | - | 15, 16 | 1658 | ||
AK122431.1 | - | 46, 47 | 5678 | |||
BC009134.1 | - | 5, 6 | 606 | |||
BC046250.1 | - | 41, 42 | 4725 | |||
BC052947.1 | - | 41, 42 | 4737 | |||
BC057692.1 | - | 11, 12 | 1250 | |||
BC157964.1 | - | 46, 47 | 5288 | 9007289 | ||
BC158050.1 | - | 46, 47 | 5607 | 9007354 | ||
BC158127.1 | - | 46, 47 | 5151 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
