siRNA Details
Gene Symbol: Ndufs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030274.1 | NP_001025445.1 | 2 | 246 | ||
GenBank mRNA | AK148505.1 | - | 2 | 245 | ||
AK152845.1 | - | 2 | 241 | |||
BC081434.1 | - | 2 | 190 | 6412214 | ||
BU534760.1 | - | 2 | 249 |
Gene Symbol: BC002163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC002163.1 | - | 1 | 233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
