siRNA Details
Gene Symbol: Ift140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134126.3 | NP_598887.3 | 20, 21 | 3618 | ||
GenBank mRNA | AK129167.1 | - | 17, 18 | 2309 | ||
AK149331.1 | - | 21, 22 | 2883 | |||
AK163782.1 | - | 20, 21 | 3246 | |||
AK172033.1 | - | 5, 6 | 754 | |||
BC009019.1 | - | 4, 5 | 472 | |||
BC139005.1 | - | 20, 21 | 2847 | 8862027 | ||
BC145494.1 | - | 20, 21 | 2848 | |||
DQ266093.1 | - | 20, 21 | 2981 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
