siRNA Details
Gene Symbol: Lars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134137.2 | NP_598898.2 | 24 | 2589 | ||
GenBank mRNA | AK129337.1 | - | 24 | 2581 | ||
AK166827.1 | - | 24 | 2577 | |||
BC006060.1 | - | 10 | 995 | |||
BC052715.1 | - | 24 | 2589 | 6836462 |

More Information
Other Species Targeted

Gene Ontology Categories:
