siRNA Details
Gene Symbol: Cyp2c50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167875.1 | NP_001161347.1 | 2 | 321 | ||
NM_001167877.1 | NP_001161349.1 | 2 | 321 | |||
NM_134144.2 | NP_598905.2 | 2 | 321 | |||
GenBank mRNA | AK149466.1 | - | 2 | 266 | ||
AY206873.1 | - | 2 | 300 | |||
BC011222.1 | - | 2 | 304 | 4195223 | ||
BC051050.1 | - | 2 | 312 | 5052536 | ||
BY086770.1 | - | 2 | 324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
