siRNA Details
Gene Symbol: CDK20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF035013.1 | - | 2 | 358 | ||
AF113130.1 | - | 2 | 366 | |||
AK075325.1 | - | 2 | 247 | |||
AK226135.1 | - | 1 | 545 | |||
AK294193.1 | - | 2 | 208 | |||
AK296328.1 | - | 2 | 206 | |||
AK298993.1 | - | 2 | 198 | |||
BC002655.2 | - | 2 | 180 | 3605918 | ||
BF966262.1 | - | 2 | 433 | |||
BX647489.1 | - | 2 | 384 | |||
CB992644.1 | - | 2 | 464 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
