siRNA Details
Gene Symbol: Dock9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081039.1 | NP_001074508.1 | 49 | 5569 | ||
NM_001128307.1 | NP_001121779.1 | 50 | 5872 | |||
NM_001128308.1 | NP_001121780.1 | 49 | 5833 | |||
NM_134074.2 | NP_598835.2 | 48 | 5419 | |||
GenBank mRNA | AK045750.1 | - | 19 | 1851 | ||
AK122431.1 | - | 50 | 5871 | |||
AK134685.1 | - | 2 | 77 | |||
BC009134.1 | - | 7 | 757 | |||
BC046250.1 | - | 43 | 4876 | |||
BC052947.1 | - | 43 | 4888 | |||
BC057692.1 | - | 13 | 1401 | |||
BC157964.1 | - | 48 | 5439 | 9007289 | ||
BC158050.1 | - | 49 | 5764 | 9007354 | ||
BC158127.1 | - | 48 | 5302 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
