siRNA Details
Gene Symbol: Scaf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134123.3 | NP_598884.2 | 4 | 602 | ||
GenBank mRNA | AK122451.1 | - | 4 | 469 | ||
AK171078.1 | - | 5 | 409 | |||
AK207242.1 | - | 4 | 205 | |||
AK211575.1 | - | 4 | 208 | |||
AK213588.1 | - | 4 | 205 | |||
AK213688.1 | - | 4 | 205 | |||
AK215751.1 | - | 4 | 201 | |||
AK218304.1 | - | 4 | 205 | |||
BC038363.1 | - | 4 | 246 | 5371946 | ||
BC075621.1 | - | 4 | 595 | 6406441 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
