siRNA Details
Gene Symbol: Ift140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134126.3 | NP_598887.3 | 21 | 3660 | ||
GenBank mRNA | AK129167.1 | - | 18 | 2351 | ||
AK149331.1 | - | 22 | 2925 | |||
AK163782.1 | - | 21 | 3288 | |||
AK172033.1 | - | 6 | 796 | |||
BC009019.1 | - | 5 | 514 | |||
BC139005.1 | - | 21 | 2889 | 8862027 | ||
BC145494.1 | - | 21 | 2890 | |||
DQ266093.1 | - | 21 | 3023 |

More Information
Other Species Targeted

Gene Ontology Categories:
