siRNA Details
Gene Symbol: AI837181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256515.1 | NP_001243444.1 | 2 | 698 | ||
NM_134149.2 | NP_598910.2 | 2 | 684 | |||
GenBank mRNA | AK077296.1 | - | 1 | 420 | ||
BC017542.1 | - | 2 | 565 | 4526963 | ||
BC024516.1 | - | 2 | 685 | 4984074 | ||
U64690.1 | - | 2 | 563 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001024233, NM_001243534, CK483724, FQ233132, BC093387.
Gene Ontology Categories:
