siRNA Details
Gene Symbol: Yars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK195910.1 | - | 1, 2 | 66 | ||
AK200644.1 | - | 1, 2 | 66 | |||
AK205652.1 | - | 1, 2 | 66 | |||
BC013552.1 | - | 2, 3 | 364 | 4236514 | ||
BC022143.1 | - | 2, 3 | 328 | 5101324 | ||
BC026615.1 | - | 2, 3 | 340 | 4976999 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
