siRNA Details
Gene Symbol: Dock9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081039.1 | NP_001074508.1 | 52, 53 | 6181 | ||
NM_001128307.1 | NP_001121779.1 | 53, 54 | 6484 | |||
NM_001128308.1 | NP_001121780.1 | 52, 53 | 6445 | |||
NM_134074.2 | NP_598835.2 | 51, 52 | 6031 | |||
GenBank mRNA | AK045750.1 | - | 22, 23 | 2463 | ||
AK122431.1 | - | 53, 54 | 6483 | |||
AK134685.1 | - | 5, 6 | 689 | |||
BC009134.1 | - | 10, 11 | 1369 | |||
BC046250.1 | - | 46, 47 | 5488 | |||
BC052947.1 | - | 46, 47 | 5500 | |||
BC057692.1 | - | 16, 17 | 2013 | |||
BC157964.1 | - | 51, 52 | 6051 | 9007289 | ||
BC158050.1 | - | 52, 53 | 6376 | 9007354 | ||
BC158127.1 | - | 51, 52 | 5914 |

More Information
Other Species Targeted

Gene Ontology Categories:
