siRNA Details
Gene Symbol: Scaf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134123.3 | NP_598884.2 | 13 | 1919 | ||
GenBank mRNA | AK032418.1 | - | 121 | |||
AK122451.1 | - | 13 | 1786 | |||
AK171078.1 | - | 14 | 1726 | |||
BC038363.1 | - | 13 | 1563 | 5371946 | ||
BC075621.1 | - | 13 | 1912 | 6406441 | ||
CA560428.1 | - | 5 | 558 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
