siRNA Details
Gene Symbol: Ift140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134126.3 | NP_598887.3 | 27 | 4593 | ||
GenBank mRNA | AK129167.1 | - | 24 | 3284 | ||
AK172033.1 | - | 12 | 1729 | |||
BC009019.1 | - | 11 | 1447 | |||
BC139005.1 | - | 27 | 3822 | 8862027 | ||
BC145494.1 | - | 27 | 3823 | |||
CX221628.1 | - | 2 | 225 | |||
DQ266093.1 | - | 27 | 3956 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_002727728, XM_006246055, XM_006246054.
Gene Ontology Categories:
