siRNA Details
Gene Symbol: Lars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134137.2 | NP_598898.2 | 28, 29 | 3110 | ||
GenBank mRNA | AK129337.1 | - | 28, 29 | 3102 | ||
AK166827.1 | - | 28, 29 | 3098 | |||
BC006060.1 | - | 14, 15 | 1516 | |||
BC052715.1 | - | 28, 29 | 3110 | 6836462 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
