siRNA Details
Gene Symbol: Yars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134151.4 | NP_598912.4 | 5 | 795 | ||
GenBank mRNA | AK028785.1 | - | 5 | 657 | ||
AK043837.1 | - | 5 | 719 | |||
AK139579.1 | - | 5 | 640 | |||
AK145356.1 | - | 5 | 688 | |||
AK151880.1 | - | 5 | 647 | |||
AK169708.1 | - | 5 | 571 | |||
AK177997.1 | - | 1 | 74 | |||
BC013552.1 | - | 5 | 726 | 4236514 | ||
BC022143.1 | - | 5 | 690 | 5101324 | ||
BC026615.1 | - | 5 | 702 | 4976999 |

More Information
Other Species Targeted

Gene Ontology Categories:
