siRNA Details
Gene Symbol: Vmn1r206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134216.1 | NP_598977.1 | 1 | 189 | ||
GenBank mRNA | BC132625.1 | - | 1 | 289 | 40130902 | |
BC138067.1 | - | 1 | 289 | 8861086 |
Gene Symbol: Vmn1r191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145849.1 | NP_665848.1 | 1 | 189 |
Gene Symbol: Vmn1r209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013787.1 | NP_001013809.1 | 1 | 189 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
