siRNA Details
Gene Symbol: Ccdc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138603.3 | NP_613069.3 | 14 | 1704 | ||
GenBank mRNA | AF229637.1 | - | 14 | 1645 | ||
AK010536.1 | - | 14 | 1622 | |||
BC011195.1 | - | 14 | 1609 | 4188459 | ||
BC057686.1 | - | 5 | 1754 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
